2,4-Difluoro-N-(4-fluorobenzyl)aniline, 97%

H56638.MDEA 85.7 EUR
ALFAH56638.MD ALFAH56638.03
2,4-Difluoro-N-(4-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1019513-68-8
Molecular Formula: C13H10F3N
Formula Weight: 237.22
MDL No.: MFCD11145793
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR