2,5-Difluoro-N-(2-fluorobenzyl)aniline, 97%

H57147.03EA 144 EUR
ALFAH57147.03 ALFAH57147.MD
2,5-Difluoro-N-(2-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1019510-60-1
Molecular Formula: C13H10F3N
Formula Weight: 237.22
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR