N-(3-Fluorobenzyl)-3,4-dimethoxyaniline, 97%

H57180.MDEA 17.9 EUR
ALFAH57180.MD ALFAH57180.03
N-(3-Fluorobenzyl)-3,4-dimethoxyaniline, 97%
Purity: 97%
CAS No.: 1019488-75-5
Molecular Formula: C15H16FNO2
Formula Weight: 261.29
MDL No.: MFCD12623894
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR