4,7-Dibrom-3-hydroxy-2-naphthoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL638528-5GEA 36.4 EUR
SIAL638528-5G
4,7-Dibrom-3-hydroxy-2-naphthoesäure, Sigma-Aldrich®
4,7-Dibrom-3-hydroxy-2-naphthoesäure
Formel: C₁₁H₆Br₂O₃
Molekulargewicht: 345,98
MDL Number: MFCD00046367
CAS-Nummer: 1779-10-8
EINECS: 217-214-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR