2-(Bromacetyl)naphthalin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL105120-5GEA 31.8 EUR
SIAL105120-5G
2-(Bromacetyl)naphthalin, Sigma-Aldrich®
2-(Bromacetyl)naphthalin
Formel: C₁₂H₉BrO
Molekulargewicht: 249,11
MDL Number: MFCD00004109
CAS-Nummer: 613-54-7
EINECS: 210-348-7
UN-Nummer: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR