2-Chlor-6-methylnicotinsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL357847-5GEA 60.9 EUR
SIAL357847-5G
2-Chlor-6-methylnicotinsäure, Sigma-Aldrich®
2-Chlor-6-methylnicotinsäure
Formel: C₇H₆ClNO₂
Molekulargewicht: 171,58
MDL Number: MFCD00134165
CAS-Nummer: 30529-70-5
EINECS: 250-229-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR