3,5-Lutidin-N-oxid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL555657-25GEA 316 EUR
SIAL555657-25G
3,5-Lutidin-N-oxid, Sigma-Aldrich®
3,5-Lutidin-N-oxid
Formel: C₇H₉NO
Molekulargewicht: 123,1525 g/mol
MDL Number: MFCD00126839
CAS-Nummer: 3718-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR