5-Brom-2-thiophencarbonitril, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ200
SIAL574430-1GEA
60
EUR
SIAL574430-1G
5-Brom-2-thiophencarbonitril, Sigma-Aldrich®
5-Brom-2-thiophencarbonitril
Formel:
C₅H₂BrNS Molekulargewicht: 188,0500 g/mol |
MDL Number:
MFCD04974080 CAS-Nummer: 2160-62-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...