5-Nitro-2-thiophencarbonitril, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
SIAL07517-1G-FEA
51.8
EUR
SIAL07517-1G-F
5-Nitro-2-thiophencarbonitril, Sigma-Aldrich®
5-Nitro-2-thiophencarbonitril
Formel:
C₅H₂N₂O₂S Molekulargewicht: 154,15 g/mol |
MDL Number:
MFCD00052401 CAS-Nummer: 16689-02-4 UN-Nummer: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...