2-Nitro-4-thiophencarbonitril

Lieferant: Thermo Scientific

DFP00020.250 DFP00020DA
MAYBDFP00020.250EA 0 EUR
MAYBDFP00020.250 MAYBDFP00020DA
2-Nitro-4-thiophencarbonitril
2-Nitro-4-thiophencarbonitril
Formel: C₅H₂N₂O₂S
Molekulargewicht: 154,15 g/mol
MDL Number: MFCD00052580
CAS-Nummer: 42137-23-5
UN-Nummer: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR