Thiophen-2-methylamin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ200
SIAL220884-5GEA
84
EUR
SIAL220884-5G
Thiophen-2-methylamin, Sigma-Aldrich®
Thiophen-2-methylamin
Formel:
C₅H₇NS Molekulargewicht: 113,19 g/mol |
MDL Number:
MFCD00005460 CAS-Nummer: 27757-85-3 EINECS: 248-639-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...