2,1,3-Benzothiadiazol-4-carbaldehyd

MAYBCC09104DAEA 0 EUR
MAYBCC09104DA MAYBCC09104CB
2,1,3-Benzothiadiazol-4-carbaldehyd
2,1,3-Benzothiadiazol-4-carbaldehyd
Formel: C₇H₄N₂OS
CAS-Nummer: 5170-68-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR