2,1,3-Benzothiadiazol-5-carbonsäure

ABCRCC09201.0001EA 0 EUR
ABCRCC09201.0001 MAYBCC09201CB MAYBCC09201DA
2,1,3-Benzothiadiazol-5-carbonsäure
2,1,3-Benzothiadiazole-5-carboxylic acid
Formel: C₇H₄N₂O₂S
MDL Number: MFCD01647555
CAS-Nummer: 16405-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR