4-Iod-2,1,3-benzothiadiazol

MAYBCC09111DAEA 0 EUR
MAYBCC09111DA MAYBCC09111CB
4-Iod-2,1,3-benzothiadiazol
4-Iodo-2,1,3-benzothiadiazole
Formel: C₆H₃IN₂S
MDL Number: MFCD02681904
CAS-Nummer: 352018-95-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR