Diisopropylphthalat, Sigma-Aldrich®

Lieferant: Merck

SIAL80137-50MLEA 186 EUR
SIAL80137-50ML
Diisopropylphthalat, Sigma-Aldrich®
Diisopropylphthalat
Formel: C₁₄H₁₈O₄
Molekulargewicht: 250,2903 g/mol
MDL Number: MFCD00053717
CAS-Nummer: 605-45-8
EINECS: 210-086-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR