Chlorbis(4-methylphenyl)phosphin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL695815-1GEA
94
EUR
SIAL695815-1G
Chlorbis(4-methylphenyl)phosphin, Sigma-Aldrich®
Chlorbis(4-methylphenyl)phosphin
Formel:
C₁₄H₁₄ClP Molekulargewicht: 248,69 |
MDL Number:
MFCD01630844 CAS-Nummer: 1019-71-2 UN-Nummer: 1760 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...