4'-Propylacetanilid
Lieferant: Thermo Scientific
MAYBBTB04635.50EA
0
EUR
MAYBBTB04635.50
4'-Propylacetanilid
4'-Propylacetanilid
Formel:
C₁₁H₁₅NO |
MDL Number:
MFCD00275765 CAS-Nummer: 20330-99-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...