2-Fluoro-6-methylaniline, 98%

H55853.03EA 105 EUR
ALFAH55853.03 ALFAH55853.06 ALFAH55853.MD
2-Fluoro-6-methylaniline, 98%
Purity: 98%
CAS No.: 443-89-0
Molecular Formula: C7H8FN
Formula Weight: 125.15
Storage Temperature: 2°C to 8°C
Flash Point: 76°(169°F)
Refractive Index: 1.5360
MDL No.: MFCD06658252
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR