6-Fluor-o-toluidin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL686700-1GEA 93.6 EUR
SIAL686700-1G
6-Fluor-o-toluidin, Sigma-Aldrich®
6-Fluor-o-toluidin
Formel: C₇H₈FN
MDL Number: MFCD06658252
CAS-Nummer: 443-89-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR