2-Allylanisol, Sigma-Aldrich®

Lieferant: Merck

SIAL706760-5GEA 100 EUR
SIAL706760-5G
2-Allylanisol, Sigma-Aldrich®
2-Allylanisol
Formel: C₁₀H₁₂O
MDL Number: MFCD00272573
CAS-Nummer: 3698-28-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR