3-Ethoxybenzolthiol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

SIAL549231-1GEA 75 EUR
SIAL549231-1G SIAL549231-5G
3-Ethoxybenzolthiol, Sigma-Aldrich®
3-Ethoxythiophenol
Formel: C₈H₁₀OS
MDL Number: MFCD03094660
CAS-Nummer: 86704-82-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR