3-Methyl-1,4-pentadien, Sigma-Aldrich®

Lieferant: Merck

SIAL68287-5MLEA 101 EUR
SIAL68287-5ML
3-Methyl-1,4-pentadien, Sigma-Aldrich®
3-Methyl-1,4-pentadiene
Formel: C₆H₁₀
MDL Number: MFCD00008634
CAS-Nummer: 1115-08-8
EINECS: 214-221-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR