2,5-Dichlor-3-methylthiophen, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIAL668362-1GEA 29.2 EUR
SIAL668362-1G
2,5-Dichlor-3-methylthiophen, Sigma-Aldrich®
2,5-Dichloro-3-methylthiophene
Formel: C₅H₄Cl₂S
Molekulargewicht: 167,06
MDL Number: MFCD00022490
CAS-Nummer: 17249-90-0
UN-Nummer: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR