2-Fluor-4-iodbenzonitril

Lieferant: Apollo Scientific

APOSPC2726-5GEA 81.4 EUR
APOSPC2726-5G
2-Fluor-4-iodbenzonitril
2-Fluor-4-iodbenzonitril
Formel: C₇H₃FIN
MDL Number: MFCD03094172
CAS-Nummer: 137553-42-5
UN-Nummer: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR