2-Methoxyethylchlorformiat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIAL592293-5MLEA 55.5 EUR
SIAL592293-5ML SIAL592293-25ML
2-Methoxyethylchlorformiat, Sigma-Aldrich®
2-Methoxyethylchlorformiat
Formel: C₄H₇ClO₃
Molekulargewicht: 138,55
MDL Number: MFCD00058932
CAS-Nummer: 628-12-6
EINECS: 211-026-9
UN-Nummer: 2742
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR