5-Formylsalicylsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIALF17601-5GEA 140 EUR
SIALF17601-5G
5-Formylsalicylsäure, Sigma-Aldrich®
5-Formylsalicylsäure
Formel: C₈H₆O₄
MDL Number: MFCD00006945
CAS-Nummer: 616-76-2
EINECS: 210-492-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR