3-Ethoxysalicylaldehyd, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL160989-50GEA
58.9
EUR
SIAL160989-50G
3-Ethoxysalicylaldehyd, Sigma-Aldrich®
3-Ethoxysalicylaldehyd
Formel:
C₉H₁₀O₃ Molekulargewicht: 166,18 |
MDL Number:
MFCD00003323 CAS-Nummer: 492-88-6 EINECS: 207-765-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...