2-Brom-3-nitrotoluol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL381799-5GEA 77.7 EUR
SIAL381799-5G
2-Brom-3-nitrotoluol, Sigma-Aldrich®
2-Brom-3-nitrotoluol
Formel: C₇H₆BrNO₂
MDL Number: MFCD00134555
CAS-Nummer: 41085-43-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR