Methyl-5-chlorsalicylat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL165018-100GEA
160
EUR
SIAL165018-100G
Methyl-5-chlorsalicylat, Sigma-Aldrich®
Methyl-5-chlorsalicylat
Formel:
C₈H₇ClO₃ Molekulargewicht: 186,60 |
MDL Number:
MFCD00016461 CAS-Nummer: 4068-78-4 EINECS: 223-778-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...