Glycidisopropylether, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL291463-250MLEA 123 EUR
SIAL291463-250ML
Glycidisopropylether, Sigma-Aldrich®
Glycidisopropylether
Formel: C₆H₁₂O₂
MDL Number: MFCD00005139
CAS-Nummer: 4016-14-2
EINECS: 223-672-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR