4'-Nitro-2'-(trifluormethyl)acetanilid
Lieferant: Thermo Scientific
MAYBTL00253.1EA
0
EUR
MAYBTL00253.1
4'-Nitro-2'-(trifluormethyl)acetanilid
4'-Nitro-2'-(trifluormethyl)acetanilid
Formel:
C₉H₇F₃N₂O₃ Molekulargewicht: 248,16 g/mol |
MDL Number:
MFCD00052846 CAS-Nummer: 395-68-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...