4-tert-Butyl-2,6-dinitroanisol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL565687-25GEA 20.6 EUR
SIAL565687-25G
4-tert-Butyl-2,6-dinitroanisol, Sigma-Aldrich®
4-tert-Butyl-2,6-dinitroanisol
Formel: C₁₁H₁₄N₂O₅
MDL Number: MFCD00185596
CAS-Nummer: 77055-30-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR