1-Benzyl-4-hydroxypiperidin, Sigma-Aldrich®

Lieferant: Merck

SIAL152986-25GEA 91.7 EUR
SIAL152986-25G
1-Benzyl-4-hydroxypiperidin, Sigma-Aldrich®
1-Benzyl-4-hydroxypiperidin
Formel: C₁₂H₁₇NO
Molekulargewicht: 191,27 g/mol
MDL Number: MFCD00006503
CAS-Nummer: 4727-72-4
EINECS: 225-226-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR