2-Amino-N-isopropylbenzamid
Lieferant: EHRENSTORFER GMBH
EHERC10280000EA
110
EUR
EHERC10280000
2-Amino-N-isopropylbenzamid
2-Amino-N-isopropylbenzamid
Formel:
C₁₀H₁₄N₂O |
MDL Number:
MFCD00143620 CAS-Nummer: 30391-89-0 EINECS: 250-175-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...