(R)-(-)-1-Benzyl-3-hydroxypiperidin ≥97%

Lieferant: Thermo Scientific
Achtung

H30995.MDEA 46.1 EUR
H30995.MD H30995.03
(R)-(-)-1-Benzyl-3-hydroxypiperidin ≥97%
(R)-(-)-1-Benzyl-3-hydroxypiperidin
Formel: C₁₂H₁₇NO
Molekulargewicht: 191,27 g/mol
MDL Number: MFCD00274307
CAS-Nummer: 91599-81-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR