4',5'-Dichlor-2'-nitroacetanilid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL641715-5GEA
120
EUR
SIAL641715-5G
4',5'-Dichlor-2'-nitroacetanilid, Sigma-Aldrich®
4',5'-Dichlor-2'-nitroacetanilid
Formel:
C₈H₆Cl₂N₂O₃ Molekulargewicht: 249,0508 g/mol |
MDL Number:
MFCD00024301 CAS-Nummer: 5462-30-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...