4-Amino-3-chlor-5-nitrobenzotrifluorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL656038-5GEA 84.7 EUR
SIAL656038-5G SIAL656038-1G
4-Amino-3-chlor-5-nitrobenzotrifluorid, Sigma-Aldrich®
4-Amino-3-chlor-5-nitrobenzotrifluorid
Formel: C₇H₄ClF₃N₂O₂
Molekulargewicht: 240,58
MDL Number: MFCD00042153
CAS-Nummer: 57729-79-0
UN-Nummer: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR