Sicherheitsschuhe für Damen, zum Schnüren, Elena

Lieferant: Jallatte
JALCJJD23/41EA 113 EUR
JALCJJD23/41 JALCJJD23/42 JALCJJD23/37 JALCJJD23/35 JALCJJD23/38 JALCJJD23/40 JALCJJD23/36 JALCJJD23/39
Sicherheitsschuhe für Damen, zum Schnüren, Elena
Safety shoes made from Aseptane™ upper (resistant to fluid spills) and dual-density TPU sole.

  • Aseptane™ upper is water repellent, breathable, washable at 30 °C
  • Antibacterial nylon lining
  • Full piece, anti-static and anatomic integrated insole, perforated in the front part for better foot venting, cushioning effect in the heel area
  • TPU/TPU versitale dual-density sole is resistant to slipping and abrasion
  • Aérfort perforated steel toecap (200 J)
  • Non magnetic, lightweight and flexible FleXtane™ HP midsole (100% composite)

Colour: White

Suitable for food industries and wet working conditions.

Zertifizierungen: EN ISO 20345:2007; S3, SRC
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR