Filter tips, GentleFit™ Dual-Shot, Axygen®
Lieferant: Corning Life Sciences
Corning® Axygen®
613-2309EA 142 EUR
613-2309 613-2312 613-2311
Filter tips, GentleFit™ Dual-Shot, Axygen®
Pipettenspitzen
These filter barrier tips are ideal for DNA amplification and microbiological applications where trace contamination can completely invalidate results.
  • PP body and TPE top, sterile
  • RNase, DNase, and endotoxin free
  • Tips create a leak proof seal: Minimal force required to load and eject tip
  • Reduced wear on pipettors – giving them a longer life
  • Ergonomic: Soft tip allows for less pressure to create an optimal seal
  • Top collar color-coded by volume