2-Amino-4-(2-pyridyl)thiazol 97%
Lieferant: Thermo Scientific
|
Achtung
|
H58554.04EA
363
EUR
H58554.04
2-Amino-4-(2-pyridyl)thiazol 97%
2-Amino-4-(2-pyridyl)thiazole
Formel:
C₈H₇N₃S Molekulargewicht: 177,22 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00460417 CAS-Nummer: 30235-26-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...