Methyl-3-hydroxyhexanoat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ300

SIALW350818-1KG-KEA 774 EUR
SIALW350818-1KG-K
Methyl-3-hydroxyhexanoat, Sigma-Aldrich®
Methyl-3-hydroxyhexanoat
Formel: C₇H₁₄O₃
MDL Number: MFCD00083583
CAS-Nummer: 21188-58-9
EINECS: 244-261-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR