3-Brom-5-(trifluormethyl)phenylboronsäurepinakolester ≥97%
H51713.03EA
259
EUR
H51713.03
3-Brom-5-(trifluormethyl)phenylboronsäurepinakolester ≥97%
3-Brom-5-(trifluormethyl)phenylboronsäurepinakolester
Formel:
C₁₃H₁₅BBrF₃O₂ Molekulargewicht: 350,97 g/mol Schmelzpunkt: 50…52 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD12405333 CAS-Nummer: 479411-92-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...