Furfuryldisulfid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL264768-1GEA 29.9 EUR
SIAL264768-1G
Furfuryldisulfid, Sigma-Aldrich®
Furfuryldisulfid
Formel: C₁₀H₁₀O₂S₂
MDL Number: MFCD00010082
CAS-Nummer: 4437-20-1
EINECS: 224-649-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR