1,3,5-Trithian, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIALT88404-100GEA 220 EUR
SIALT88404-100G
1,3,5-Trithian, Sigma-Aldrich®
1,3,5-Trithiane
Formel: C₃H₆S₃
MDL Number: MFCD00006653
CAS-Nummer: 291-21-4
EINECS: 206-029-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR