2-Amino-3-formylchromon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL402028-1GEA 146 EUR
SIAL402028-1G
2-Amino-3-formylchromon, Sigma-Aldrich®
2-Amino-3-formylchromon
Formel: C₁₀H₇NO₃
Molekulargewicht: 189,1675 g/mol
MDL Number: MFCD00191735
CAS-Nummer: 61424-76-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR