6-Fluor-3-formylchromon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL543624-1GEA 45 EUR
SIAL543624-1G
6-Fluor-3-formylchromon, Sigma-Aldrich®
6-Fluor-3-formylchromon
Formel: C₁₀H₅FO₃
Molekulargewicht: 192,15 g/mol
MDL Number: MFCD00139060
CAS-Nummer: 69155-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR