Tetrabenzylpyrophosphat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ200
SIAL418633-1GEA
109
EUR
SIAL418633-1G
Tetrabenzylpyrophosphat, Sigma-Aldrich®
Tetrabenzylpyrophosphat
Formel:
C₂₈H₂₈O₇P₂ Molekulargewicht: 538,48 g/mol |
MDL Number:
MFCD00051941 CAS-Nummer: 990-91-0 UN-Nummer: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...