5-Indolboronsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL666467-1GEA 47.3 EUR
SIAL666467-1G
5-Indolboronsäure, Sigma-Aldrich®
5-Indoleboronic acid
Formel: C₈H₈BNO₂
Molekulargewicht: 160,97 g/mol
MDL Number: MFCD01319013
CAS-Nummer: 144104-59-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR