1-(Phenylsulfonyl)-5-indolboronsäure

Lieferant: Molekula

MOLEM27606540EA 0 EUR
MOLEM27606540
1-(Phenylsulfonyl)-5-indolboronsäure
1-(Phenylsulphonyl)-5-indoleboronic acid
Formel: C₁₄H₁₂BNO₄S
Molekulargewicht: 301,1300 g/mol
MDL Number: MFCD03840606
CAS-Nummer: 480438-51-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR