1-(Phenylsulfonyl)-6-indolboronsäure

Lieferant: Molekula

MOLEM35514231EA 0 EUR
MOLEM35514231
1-(Phenylsulfonyl)-6-indolboronsäure
1-(Phenylsulphonyl)-6-indoleboronic acid
Formel: C₁₄H₁₂BNO₄S
Molekulargewicht: 301,1254 g/mol
MDL Number: MFCD03840611
CAS-Nummer: 480438-52-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR